Assay Details
| Gene Symbol: Nrg3os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045713.1 | 1 - 2 | 64 | 186 | ||
| GenBank mRNA | AK015934.1 | - | 1 - 2 | 65 | 186 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nrg3os
Genomic Map