| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400003522.mm.1 |
NR_045713.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003522.mm.1 |
NR_045713.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003522.mm.1 |
TC1400000400.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400003526.mm.1 |
NR_045713.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003526.mm.1 |
NR_045713.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003526.mm.1 |
TC1400000400.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400003524.mm.1 |
NR_045713.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003524.mm.1 |
NR_045713.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003524.mm.1 |
TC1400000400.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400003523.mm.1 |
NR_045713.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003523.mm.1 |
NR_045713.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003523.mm.1 |
TC1400000400.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400003521.mm.1 |
NR_045713.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003521.mm.1 |
NR_045713.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003521.mm.1 |
TC1400000400.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400003519.mm.1 |
NR_045713.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003519.mm.1 |
NR_045713.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003519.mm.1 |
TC1400000400.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400003527.mm.1 |
NR_045713.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003527.mm.1 |
NR_045713.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003527.mm.1 |
TC1400000400.mm.1 |
MTA TC |