Assay Details
| Gene Symbol: Donson | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021720.1 | NP_068366.1 | 1 - 2 | 322 | 91 | |
| GenBank mRNA | AK042415.1 | - | 1 - 2 | 347 | 91 | |
| AK132269.1 | - | 1 - 2 | 346 | 91 | ||
| BC018792.1 | - | 1 - 2 | 321 | 91 | ||
| BC043316.1 | - | 1 - 2 | 322 | 5320969 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Donson
Genomic Map