Assay Details
| Gene Symbol: Donson | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021720.1 | NP_068366.1 | 7 - 8 | 1150 | 91 | |
| GenBank mRNA | AF193608.1 | - | 4 - 5 | 441 | 91 | |
| AK042415.1 | - | 6 - 7 | 1297 | 91 | ||
| AK132269.1 | - | 7 - 8 | 1174 | 91 | ||
| BC018792.1 | - | 6 - 7 | 970 | 91 | ||
| BC043316.1 | - | 7 - 8 | 1150 | 5320969 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Donson
Genomic Map