Assay Details
| Gene Symbol: 9230114K14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079034.1 | - | 231 | 198 | ||
| AK163067.1 | - | 1 - 2 | 268 | 198 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

9230114K14Rik| Gene Symbol: 9230114K14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079034.1 | - | 231 | 198 | ||
| AK163067.1 | - | 1 - 2 | 268 | 198 | ||
Set Membership: |
|