Assay Details
Gene Symbol: Gm11872 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK029221.1 | - | 1 - 2 | 190 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
