Assay Details
| Gene Symbol: Gm15997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045423.1 | 1 - 2 | 118 | 99 | ||
| GenBank mRNA | AK043895.1 | - | 1 - 2 | 119 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WD40 repeat domain 95 | Wdr95 | 4930434E21Rik | Anti-sense | Chr. 5 - 149528679 - 149611894 on NCBI Build GRCm38 | |
| Downstream | heat shock 105kDa/110kDa protein 1 | Hsph1 |
105kDa, AI790491, Hsp105, Hsp110, hsp-E7I, hsp110/105 |
78.8kb | Sense | Chr. 5 - 149616843 - 149637155 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm15997
Genomic Map