Assay Details
| Gene Symbol: Gm15997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045423.1 | 3 - 4 | 433 | 74 | ||
| GenBank mRNA | AK043895.1 | - | 3 - 4 | 434 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | testis expressed 26 | Tex26 | 4930588N13Rik | -22.5kb | Anti-sense | Chr. 5 - 149439517 - 149470620 on NCBI Build GRCm38 |
| Downstream | WD40 repeat domain 95 | Wdr95 | 4930434E21Rik | 33.9kb | Anti-sense | Chr. 5 - 149528679 - 149611894 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm15997
Genomic Map