Assay Details
| Gene Symbol: Gm15997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045423.1 | 4 - 5 | 625 | 62 | ||
| GenBank mRNA | AK043895.1 | - | 4 - 5 | 626 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | testis expressed 26 | Tex26 | 4930588N13Rik | -18.4kb | Anti-sense | Chr. 5 - 149439517 - 149470620 on NCBI Build GRCm38 |
| Downstream | WD40 repeat domain 95 | Wdr95 | 4930434E21Rik | 35.3kb | Anti-sense | Chr. 5 - 149528679 - 149611894 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 440990 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 254580 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 242251 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801686 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 406874 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 238637 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 833792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235261 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 382074 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 377994 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559381 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510683 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10622 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 452403 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 503787 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608953 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 519081 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 384949 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 327372 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 498875 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 875105 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 139332 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 168704 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 70017 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P101045 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P101045 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01250175 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01250169 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101881 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P101045 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01250169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04424463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04363092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04554360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04328210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04478064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04922792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185454 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04328207 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04328217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185578 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04633206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185572 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04328208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04291682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04424455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04613772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04424451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04363089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04424466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04335122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185479 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185550 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04424449 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185516 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04922797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04613786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04979021 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04477249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04613781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04922794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04424459 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04613777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04191090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04613787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185561 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04730301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04424458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04424457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04328214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185506 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24185514 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR0500033565.mm.1 | TC0500003658.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500033565.mm.1 | NR_045423.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500033565.mm.1 | NR_045423.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 468772 | - | - |

Gm15997
Genomic Map