Assay Details
| Gene Symbol: Gm14342 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK045142.1 | - | 95 | 66 | |||
| DQ860415.1 | - | LIT3343 HAR1R |
143 | 66 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
basic helix-loop-helix family, member e23 |
Bhlhe23 |
A930001L02Rik, BETA4, Bhlhb4 |
-63.1kb | Sense | Chr. 2 - 180774381 - 180776900 on NCBI Build GRCm38 |
| Downstream | microRNA 124a-3 | Mir124a-3 |
Mirn124a-3, mir-124-3, mir-124a-3 |
4.4kb | Anti-sense | Chr. 2 - 180894040 - 180894107 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm14342
Genomic Map