Assay Details
Gene Symbol: Gm31343 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK042586.1 | - | 4 - 5 | 1620 | 108 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
solute carrier family 10, member 2 |
Slc10a2 |
ASBT, ISBT |
-3.3mb | Anti-sense | Chr. 8 - 5085623 - 5105232 on NCBI Build GRCm38 |
Downstream |
predicted gene, 31135 |
Gm31135 | 44.9kb | Anti-sense | Chr. 8 - 8436606 - 8484728 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
