| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400002054.mm.1 |
NM_001134465.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002054.mm.1 |
NM_001285466.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002054.mm.1 |
NM_001285467.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002054.mm.1 |
NM_145969.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002054.mm.1 |
NM_145969.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002054.mm.1 |
TC1400000261.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400002057.mm.1 |
NM_001134465.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002057.mm.1 |
NM_001285466.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002057.mm.1 |
NM_001285467.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002057.mm.1 |
NM_145969.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002057.mm.1 |
NM_145969.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002057.mm.1 |
TC1400000261.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400002059.mm.1 |
NM_001134465.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002059.mm.1 |
NM_001285466.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002059.mm.1 |
NM_001285467.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002059.mm.1 |
TC1400000261.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400002055.mm.1 |
NM_001134465.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002055.mm.1 |
NM_001285466.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002055.mm.1 |
NM_001285467.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002055.mm.1 |
NM_145969.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002055.mm.1 |
NM_145969.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002055.mm.1 |
TC1400000261.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400002053.mm.1 |
NM_001134465.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002053.mm.1 |
NM_001285466.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002053.mm.1 |
NM_001285467.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002053.mm.1 |
NM_145969.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002053.mm.1 |
NM_145969.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002053.mm.1 |
TC1400000261.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400002056.mm.1 |
NM_001134465.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002056.mm.1 |
NM_001285466.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002056.mm.1 |
NM_001285467.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002056.mm.1 |
NM_145969.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002056.mm.1 |
NM_145969.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400002056.mm.1 |
TC1400000261.mm.1 |
MTA TC |