Assay Details
| Gene Symbol: Bod1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081422.2 | NP_001074891.1 | 1 - 2 | 383 | 81 | |
| GenBank mRNA | AK046296.1 | - | 1 - 2 | 356 | 81 | |
| AK053690.1 | - | 1 - 2 | 361 | 81 | ||
| AK077824.1 | - | 1 - 2 | 384 | 81 | ||
| AK079548.1 | - | 1 - 2 | 367 | 81 | ||
| AK134590.1 | - | 1 - 2 | 361 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bod1l
Genomic Map