Assay Details
| Gene Symbol: Bod1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081422.2 | NP_001074891.1 | 2 - 3 | 513 | 107 | |
| GenBank mRNA | AK046296.1 | - | 2 - 3 | 486 | 107 | |
| AK053690.1 | - | 2 - 3 | 491 | 107 | ||
| AK079548.1 | - | 2 - 3 | 497 | 107 | ||
| AK134590.1 | - | 2 - 3 | 491 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bod1l
Genomic Map