Assay Details
| Gene Symbol: Bod1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081422.2 | NP_001074891.1 | 3 - 4 | 700 | 88 | |
| GenBank mRNA | AK046296.1 | - | 3 - 4 | 673 | 88 | |
| AK053690.1 | - | 3 - 4 | 678 | 88 | ||
| AK079548.1 | - | 3 - 4 | 684 | 88 | ||
| AK134590.1 | - | 3 - 4 | 678 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bod1l
Genomic Map