Assay Details
| Gene Symbol: Bod1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081422.2 | NP_001074891.1 | 8 - 9 | 1882 | 150 | |
| GenBank mRNA | AK077824.1 | - | 8 - 9 | 1883 | 150 | |
| AK134590.1 | - | 8 - 9 | 1860 | 150 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bod1l
Genomic Map