Assay Details
| Gene Symbol: Bod1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081422.2 | NP_001074891.1 | 23 - 24 | 8748 | 98 | |
| GenBank mRNA | AF013969.1 | - | 14 - 15 | 4207 | 98 | |
| AK122495.1 | - | 14 - 15 | 3892 | 98 | ||
| BC024569.1 | - | 2 - 3 | 98 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bod1l
Genomic Map