Assay Details
| Gene Symbol: Bod1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081422.2 | NP_001074891.1 | 10 - 11 | 7907 | 106 | |
| GenBank mRNA | AF013969.1 | - | 1 - 2 | 3366 | 106 | |
| AK122495.1 | - | 1 - 2 | 3051 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bod1l
Genomic Map