Assay Details
| Gene Symbol: Bod1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081422.2 | NP_001074891.1 | 15 - 16 | 8229 | 81 | |
| GenBank mRNA | AF013969.1 | - | 6 - 7 | 3688 | 81 | |
| AK122495.1 | - | 6 - 7 | 3373 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bod1l
Genomic Map