Assay Details
| Gene Symbol: Bod1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081422.2 | NP_001074891.1 | 16 - 17 | 8307 | 105 | |
| GenBank mRNA | AF013969.1 | - | 7 - 8 | 3766 | 105 | |
| AK122495.1 | - | 7 - 8 | 3451 | 105 | ||
More Information
Set Membership: |
|

Bod1l
Genomic Map