Assay Details
| Gene Symbol: Bod1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081422.2 | NP_001074891.1 | 18 - 19 | 8469 | 110 | |
| GenBank mRNA | AF013969.1 | - | 9 - 10 | 3928 | 110 | |
| AK122495.1 | - | 9 - 10 | 3613 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bod1l
Genomic Map