Assay Details
| Gene Symbol: Proz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC024104.1 | - | 2 - 3 | 485 | 5136392 | 108 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC024104.1 transcripts.
Set Membership: |
|

Proz
Genomic Map