Assay Details
| Gene Symbol: Mapk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016700.4 | NP_057909.1 | 10 - 11 | 1248 | 71 | |
| GenBank mRNA | AB005663.1 | - | 9 - 10 | 1081 | 71 | |
| AB206575.1 | - | 9 - 10 | 1023 | 71 | ||
| AB219977.1 | - | 9 - 10 | 1057 | 71 | ||
| AK043102.1 | - | 10 - 11 | 1248 | 71 | ||
| AK139498.1 | - | 10 - 11 | 1236 | 71 | ||
| BC053027.1 | - | 10 - 11 | 1218 | 5694771 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mapk8
Genomic Map