Assay Details
| Gene Symbol: Mapk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016700.4 | NP_057909.1 | 2 - 3 | 313 | 85 | |
| GenBank mRNA | AB005663.1 | - | 1 - 2 | 146 | 85 | |
| AB206575.1 | - | 1 - 2 | 122 | 85 | ||
| AB219977.1 | - | 1 - 2 | 122 | 85 | ||
| AK030767.1 | - | 2 - 3 | 315 | 85 | ||
| AK032486.1 | - | 2 - 3 | 311 | 85 | ||
| AK043102.1 | - | 2 - 3 | 313 | 85 | ||
| AK085745.1 | - | 2 - 3 | 313 | 85 | ||
| BB619635.1 | - | 2 - 3 | 315 | 85 | ||
| BC053027.1 | - | 2 - 3 | 283 | 5694771 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mapk8
Genomic Map