Assay Details
| Gene Symbol: Mapk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB219977.1 | - | 6 - 7 | 680 | 85 | |
| AK139498.1 | - | 7 - 8 | 859 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mapk8| Gene Symbol: Mapk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB219977.1 | - | 6 - 7 | 680 | 85 | |
| AK139498.1 | - | 7 - 8 | 859 | 85 | ||
Set Membership: |
|