Assay Details
| Gene Symbol: Fam160a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172682.3 | NP_766270.2 | 7 - 8 | 1461 | 77 | |
| GenBank mRNA | AK036881.1 | - | 4 - 5 | 447 | 77 | |
| BC094511.1 | - | 5 - 6 | 672 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam160a1
Genomic Map