Assay Details
| Gene Symbol: B3galt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033149.3 | NP_149161.1 | 1 - 2 | 49 | 133 | |
| GenBank mRNA | BC057887.1 | - | 1 - 2 | 49 | 4983830 | 133 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC057887.1, NM_033149.3 transcripts.
Set Membership: |
|

B3galt5
Genomic Map