Assay Details
| Gene Symbol: Rgs19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291206.1 | NP_001278135.1 | 1 - 2 | 445 | 80 | |
| GenBank mRNA | AF535869.1 | - | 1 - 2 | 446 | 80 | |
| BG085760.2 | - | 1 - 2 | 247 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF535869.1, NM_001291206.1 transcripts.
Set Membership: |
|

Rgs19
Genomic Map