Assay Details
| Gene Symbol: Gm10785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040389.1 | 1 - 2 | 386 | 71 | ||
| GenBank mRNA | AK038992.1 | - | 1 - 2 | 373 | 71 | |
| AK160609.1 | - | 1 - 2 | 387 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK160609.1 transcripts.
Set Membership: |
|

Gm10785
Genomic Map