Assay Details
| Gene Symbol: 4930513D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045416.1 | 2 - 3 | 201 | 88 | ||
| GenBank mRNA | AK015779.1 | - | 2 - 3 | 201 | 88 | |
| AK132902.1 | - | 2 - 3 | 160 | 88 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cytokine-dependent hematopoietic cell linker | Clnk | MIST | -632.8kb | Sense | Chr. 5 - 38705588 - 38876858 on NCBI Build GRCm38 |
| Downstream | heparan sulfate (glucosamine) 3-O-sulfotransferase 1 | Hs3st1 |
3-Ost, D5Wsu110e, Hsg3ost |
75.0kb | Sense | Chr. 5 - 39613935 - 39644631 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930513D17Rik
Genomic Map