Assay Details
| Gene Symbol: 4930513D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045416.1 | 3 - 4 | 376 | 107 | ||
| GenBank mRNA | AK015779.1 | - | 3 - 4 | 376 | 107 | |
| AK132902.1 | - | 3 - 4 | 335 | 107 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cytokine-dependent hematopoietic cell linker | Clnk | MIST | -600.1kb | Sense | Chr. 5 - 38705588 - 38876858 on NCBI Build GRCm38 |
| Downstream | heparan sulfate (glucosamine) 3-O-sulfotransferase 1 | Hs3st1 |
3-Ost, D5Wsu110e, Hsg3ost |
104.1kb | Sense | Chr. 5 - 39613935 - 39644631 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506581 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 564255 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 159033 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 518685 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 941112 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 165178 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 362621 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61055 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 817872 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 855884 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 295797 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 394476 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939598 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 220979 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 637767 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 883164 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 153196 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 659621 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 505553 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706680 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409564 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 595302 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 565611 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 847056 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 261465 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 886584 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 519356 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 343835 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 98132 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204529 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 438899 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478880 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 951750 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375062 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206052 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 486320 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329432 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 56284 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 391825 | AK076294.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215068 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16822 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 8341 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 47960 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84560 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127270 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 168371 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 85846 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 172696 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 71338 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05543298 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05543222 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05543372 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01080635 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05543348 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01080753 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05543200 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05543337 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05543261 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05543320 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05543261 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05543200 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05543337 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05543372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626971 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626848 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04894507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04708377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04574410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04149541 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04285259 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04488399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627116 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627225 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04107006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04855559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04723778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627148 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04644866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627063 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04722825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04106968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627199 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04510876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04908429 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627059 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04580616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627205 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627062 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04380654 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04296530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04296529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04510873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627172 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23627157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23626969 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR0500021169.mm.1 | TC0500002294.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500021169.mm.1 | NR_045416.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500021169.mm.1 | NR_045416.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500021172.mm.1 | TC0500002294.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500021172.mm.1 | NR_045416.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500021172.mm.1 | NR_045416.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500021171.mm.1 | TC0500002294.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500021171.mm.1 | NR_045416.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500021171.mm.1 | NR_045416.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 846816 | - | - |

4930513D17Rik
Genomic Map