Assay Details
| Gene Symbol: Gm36814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK030154.1 | - | 581 | 121 | ||
| AK132853.1 | - | 4 - 5 | 415 | 121 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 8117 | Mir8117 |
Gm27323, mmu-mir-8117 |
-274.1kb | Anti-sense | Chr. 5 - 50252762 - 50252868 on NCBI Build GRCm38 |
| Downstream |
peroxisome proliferative activated receptor, gamma, coactivator 1 alpha |
Ppargc1a |
A830037N07Rik, ENSMUSG00000079510, Gm11133, PGC-1, PGC-1v, Pgc-1alpha, Pgc1, Pgco1, Ppargc1 |
925.4kb | Sense | Chr. 5 - 51454249 - 52115958 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm36814
Genomic Map