Assay Details
| Gene Symbol: 5730480H06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045500.1 | 3 - 4 | 1281 | 124 | ||
| GenBank mRNA | AK017706.1 | - | 3 - 4 | 1282 | 124 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Kv channel interacting protein 4 | Kcnip4 |
AV032399, Calp, Calp250, KChIP4, KChIP4a |
Anti-sense | Chr. 5 - 48389503 - 49525122 on NCBI Build GRCm38 | |
| Downstream | adhesion G protein-coupled receptor A2 | Adgra2 |
3830613O22Rik, AU044632, Gpr125, Tem5-like |
1.5mb | Anti-sense | Chr. 5 - 49959951 - 50058996 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

5730480H06Rik
Genomic Map