Assay Details
| Gene Symbol: Pnisr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025669.1 | NP_079945.1 | 9 - 10 | 1724 | 80 | |
| GenBank mRNA | AK012036.1 | - | 9 - 10 | 1266 | 80 | |
| AK017507.1 | - | 9 - 10 | 1251 | 80 | ||
| AK020035.1 | - | 9 - 10 | 1257 | 80 | ||
| AK031964.1 | - | 9 - 10 | 1260 | 80 | ||
| AK077040.1 | - | 7 - 8 | 2973 | 80 | ||
| BC141031.1 | - | 9 - 10 | 1588 | 9055643 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pnisr
Genomic Map