Assay Details
Gene Symbol: Lor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008508.2 | NP_032534.2 | 1 - 2 | 47 | 91 | |
GenBank mRNA | BC026781.1 | - | 1 - 2 | 60 | 91 | |
BC058223.1 | - | 1 - 2 | 35 | 6307099 | 91 | |
CJ043484.1 | - | 1 - 2 | 47 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC058223.1, NM_008508.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
