Assay Details
| Gene Symbol: Ntrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033124.1 | NP_001028296.1 | 12 - 13 | 1526 | 61 | |
| GenBank mRNA | AK081588.1 | - | 12 - 13 | 1529 | 61 | |
| AK148691.1 | - | 5 - 6 | 435 | 61 | ||
More Information
Set Membership: |
|

Ntrk1
Genomic Map