Assay Details
| Gene Symbol: Ntrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033124.1 | NP_001028296.1 | 13 - 14 | 1658 | 91 | |
| GenBank mRNA | AK081588.1 | - | 13 - 14 | 1661 | 91 | |
| AK148691.1 | - | 6 - 7 | 567 | 91 | ||
| BC051596.1 | - | 3 - 4 | 352 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ntrk1
Genomic Map