Assay Details
| Gene Symbol: Ntrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033124.1 | NP_001028296.1 | 6 - 7 | 740 | 65 | |
| GenBank mRNA | AK081588.1 | - | 6 - 7 | 743 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ntrk1
Genomic Map