Assay Details
| Gene Symbol: Ntrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033124.1 | NP_001028296.1 | 7 - 8 | 881 | 102 | |
| GenBank mRNA | AK081588.1 | - | 7 - 8 | 884 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ntrk1
Genomic Map