Assay Details
| Gene Symbol: Ntrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033124.1 | NP_001028296.1 | 11 - 12 | 1384 | 65 | |
| GenBank mRNA | AK081588.1 | - | 11 - 12 | 1387 | 65 | |
| AK148691.1 | - | 4 - 5 | 293 | 65 | ||
More Information
Set Membership: |
|

Ntrk1
Genomic Map