Assay Details
| Gene Symbol: Slc26a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145947.2 | NP_666059.2 | 11 - 12 | 1445 | 120 | |
| GenBank mRNA | AF345194.1 | - | 10 - 11 | 1347 | 120 | |
| AK054327.1 | - | 11 - 12 | 1446 | 120 | ||
| BC026928.1 | - | 11 - 12 | 1424 | 4020760 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc26a7
Genomic Map