Assay Details
| Gene Symbol: Slc26a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145947.2 | NP_666059.2 | 14 - 15 | 1749 | 65 | |
| GenBank mRNA | AF345194.1 | - | 13 - 14 | 1651 | 65 | |
| BC026928.1 | - | 14 - 15 | 1728 | 4020760 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc26a7
Genomic Map