Assay Details
| Gene Symbol: Car8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007592.3 | NP_031618.2 | 6 - 7 | 713 | 148 | |
| GenBank mRNA | AK004896.1 | - | 6 - 7 | 714 | 148 | |
| AK004923.1 | - | 6 - 7 | 714 | 148 | ||
| BC010773.1 | - | 6 - 7 | 689 | 4216498 | 148 | |
| EU234006.1 | - | 5 - 6 | 539 | 148 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Car8
Genomic Map