Assay Details
| Gene Symbol: Pdgfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019971.2 | NP_064355.1 | 2 - 3 | 1331 | 76 | |
| GenBank mRNA | AF117608.1 | - | 2 - 3 | 507 | 76 | |
| AF266467.1 | - | 2 - 3 | 1331 | 76 | ||
| AK033734.1 | - | 2 - 3 | 1060 | 76 | ||
| AK042767.1 | - | 2 - 3 | 1078 | 76 | ||
| AK052947.1 | - | 2 - 3 | 583 | 76 | ||
| AK081347.1 | - | 3 - 4 | 383 | 76 | ||
More Information
Set Membership: |
|

Pdgfc
Genomic Map