Assay Details
| Gene Symbol: Pdgfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019971.2 | NP_064355.1 | 3 - 4 | 1519 | 72 | |
| GenBank mRNA | AF117608.1 | - | 3 - 4 | 695 | 72 | |
| AF266467.1 | - | 3 - 4 | 1519 | 72 | ||
| AF286725.1 | - | 3 - 4 | 498 | 72 | ||
| AK033734.1 | - | 3 - 4 | 1248 | 72 | ||
| AK042767.1 | - | 3 - 4 | 1266 | 72 | ||
| AK052947.1 | - | 3 - 4 | 771 | 72 | ||
| AK081347.1 | - | 4 - 5 | 571 | 72 | ||
| BC029099.1 | - | 4 - 5 | 822 | 72 | ||
| BC037696.1 | - | 3 - 4 | 706 | 4008749 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdgfc
Genomic Map