Assay Details
| Gene Symbol: Trim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271725.1 | NP_001258654.1 | 7 - 8 | 1731 | 94 | |
| NM_001271726.1 | NP_001258655.1 | 7 - 8 | 1767 | 94 | ||
| NM_001271727.1 | NP_001258656.1 | 7 - 8 | 2036 | 94 | ||
| NM_001271728.1 | NP_001258657.1 | 7 - 8 | 1834 | 94 | ||
| NM_030706.3 | NP_109631.2 | 7 - 8 | 1830 | 94 | ||
| GenBank mRNA | AB043550.1 | - | 7 - 8 | 1814 | 94 | |
| AF220017.1 | - | 7 - 8 | 1679 | 94 | ||
| AK045410.1 | - | 7 - 8 | 1827 | 94 | ||
| AK079415.1 | - | 7 - 8 | 1981 | 94 | ||
| AK141419.1 | - | 7 - 8 | 1677 | 94 | ||
| AK147275.1 | - | 7 - 8 | 1944 | 94 | ||
| AK147395.1 | - | 7 - 8 | 1825 | 94 | ||
| AK164350.1 | - | 6 - 7 | 1533 | 94 | ||
| AK220343.1 | - | 7 - 8 | 2035 | 94 | ||
| BC058668.1 | - | 6 - 7 | 2642 | 94 | ||
| BC058961.1 | - | 7 - 8 | 1834 | 6826143 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim2
Genomic Map