Assay Details
| Gene Symbol: Trim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271725.1 | NP_001258654.1 | 8 - 9 | 1895 | 74 | |
| NM_001271726.1 | NP_001258655.1 | 8 - 9 | 1931 | 74 | ||
| NM_001271727.1 | NP_001258656.1 | 8 - 9 | 2200 | 74 | ||
| NM_001271728.1 | NP_001258657.1 | 8 - 9 | 1998 | 74 | ||
| NM_030706.3 | NP_109631.2 | 8 - 9 | 1994 | 74 | ||
| GenBank mRNA | AB043550.1 | - | 8 - 9 | 1978 | 74 | |
| AF220017.1 | - | 8 - 9 | 1843 | 74 | ||
| AK045410.1 | - | 8 - 9 | 1991 | 74 | ||
| AK079415.1 | - | 8 - 9 | 2145 | 74 | ||
| AK141419.1 | - | 8 - 9 | 1841 | 74 | ||
| AK147275.1 | - | 8 - 9 | 2108 | 74 | ||
| AK147395.1 | - | 8 - 9 | 1989 | 74 | ||
| AK164350.1 | - | 7 - 8 | 1697 | 74 | ||
| AK220343.1 | - | 8 - 9 | 2199 | 74 | ||
| BC058668.1 | - | 7 - 8 | 2806 | 74 | ||
| BC058961.1 | - | 8 - 9 | 1998 | 6826143 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim2
Genomic Map