Assay Details
| Gene Symbol: Trim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271725.1 | NP_001258654.1 | 9 - 10 | 2066 | 158 | |
| NM_001271726.1 | NP_001258655.1 | 9 - 10 | 2102 | 158 | ||
| NM_001271727.1 | NP_001258656.1 | 9 - 10 | 2371 | 158 | ||
| NM_001271728.1 | NP_001258657.1 | 9 - 10 | 2169 | 158 | ||
| NM_030706.3 | NP_109631.2 | 9 - 10 | 2165 | 158 | ||
| GenBank mRNA | AB043550.1 | - | 9 - 10 | 2149 | 158 | |
| AF220017.1 | - | 9 - 10 | 2014 | 158 | ||
| AK042752.1 | - | 1 - 2 | 158 | 158 | ||
| AK045410.1 | - | 9 - 10 | 2162 | 158 | ||
| AK079415.1 | - | 9 - 10 | 2316 | 158 | ||
| AK141419.1 | - | 9 - 10 | 2012 | 158 | ||
| AK147275.1 | - | 9 - 10 | 2279 | 158 | ||
| AK147395.1 | - | 9 - 10 | 2160 | 158 | ||
| AK164350.1 | - | 8 - 9 | 1868 | 158 | ||
| BC058668.1 | - | 8 - 9 | 2977 | 158 | ||
| BC058961.1 | - | 9 - 10 | 2169 | 6826143 | 158 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim2
Genomic Map