Assay Details
| Gene Symbol: Fhdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033301.4 | NP_001028473.2 | 3 - 4 | 774 | 102 | |
| NM_001205355.1 | NP_001192284.1 | 3 - 4 | 845 | 102 | ||
| GenBank mRNA | AK031946.1 | - | 2 - 3 | 250 | 102 | |
| AK145217.1 | - | 3 - 4 | 765 | 102 | ||
| AK173244.1 | - | 3 - 4 | 845 | 102 | ||
| BC042779.1 | - | 4 - 5 | 460 | 102 | ||
| BC137632.1 | - | 3 - 4 | 814 | 8860651 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fhdc1
Genomic Map