Assay Details
| Gene Symbol: Fhdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033301.4 | NP_001028473.2 | 6 - 7 | 1067 | 81 | |
| NM_001205355.1 | NP_001192284.1 | 6 - 7 | 1138 | 81 | ||
| GenBank mRNA | BC137632.1 | - | 6 - 7 | 1107 | 8860651 | 81 |
More Information
Set Membership: |
|

Fhdc1
Genomic Map